accessibility menu, dialog, popup

UserName

H3CNH3CONCH3CH3

CAS RN 1187-03-7

IUPAC Name: 1,1,3,3-tetraethylurea
Synonyms: urea, tetraethyl-
Molecular Weight (g/mol): 172.27
Molecular Formula: C9H20N2O
InChi Key: UWHSPZZUAYSGTB-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)N(CC)CC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)