missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetraethylurea 99.0+%, TCI America™

Catalog No. T058725ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T058725ML 25 mL
T0587100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T058725ML Supplier TCI America Supplier No. T058725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1187-03-7
Molecular Formula C9H20N2O
Molecular Weight (g/mol) 172.272
MDL Number MFCD00042881
InChI Key UWHSPZZUAYSGTB-UHFFFAOYSA-N
Synonym tetraethylurea, urea, tetraethyl, n,n,n',n'-tetraethylurea, unii-w511x6rp5r, urea, 1,1,3,3-tetraethyl, urea, n,n,n',n'-tetraethyl, teu, tetraethyl-urea, urea,tetraethyl, 1,1,3,3-tetraethyl-ure
PubChem CID 14465
IUPAC Name 1,1,3,3-tetraethylurea
SMILES CCN(CC)C(=O)N(CC)CC

Specifications

Color Yellow
Boiling Point 214°C
Quantity 25 mL
Formula Weight 172.27
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetraethylurea
RTECSNumber YU2100000
TSCA Yes