accessibility menu, dialog, popup

UserName

CAS RN 118-69-4

IUPAC Name: 1,3-dichloro-2-methylbenzene
Synonyms: 2,6-dichlorotoluene
Molecular Weight (g/mol): 161.03
Molecular Formula: C7H6Cl2
InChi Key: DMEDNTFWIHCBRK-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC=C1Cl
Boiling Point 
  • (6)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (6)