missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorotoluene 99.0+%, TCI America™

Catalog No. D0422500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0422500G 500 g
D042225G 25 g
D0422100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0422500G Supplier TCI America Supplier No. D0422500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118-69-4
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000576
InChI Key DMEDNTFWIHCBRK-UHFFFAOYSA-N
Synonym 2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, toluene, 2,6-dichloro, unii-rvm09757ww, 1,3-dichloro-2-methyl-benzene, toluene,6-dichloro, pubchem3701, 2,6-dichloro-toluene, benzene,dichloromethyl, 2,6-dichloro toluene
PubChem CID 8368
IUPAC Name 1,3-dichloro-2-methylbenzene
SMILES CC1=C(C=CC=C1Cl)Cl

Specifications

Color Colorless
Boiling Point 198°C
Quantity 500 g
UN Number 3082
Formula Weight 161.03
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Dichlorotoluene
EINECSNumber (3)-0078
TSCA Yes