accessibility menu, dialog, popup

UserName

CAS RN 118-60-5

IUPAC Name: 2-ethylhexyl 2-hydroxybenzoate
Synonyms: octisalate ethyl hexyl salicylate octyl salicylate preparation sunarome O WMO octyl salicylate (substance) sunarome wmo octylsalicylate usaf DO-11 ethylhexyl salicylate
Molecular Weight (g/mol): 250.34
Molecular Formula: C15H22O3
InChi Key: FMRHJJZUHUTGKE-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)C1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Grade 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)