missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl salicylate, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012699
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012699 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012699 Supplier Merck Emd Millipore Supplier No. 521841ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2-Ethylhexyl salicylate is used as a UV filter in sunscreens.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 118-60-5
Density 1.014 g/mL (at 25°C)
Flash Point 113°C
Physical Form Neat
Linear Formula (HO)C6H4 CO2 CH2 CH(C2H5)(CH2)3 CH3
MDL Number MFCD00053300
Boiling Point 189°C to 190°C (21 mmHg)
Synonym 2-Ethylhexyl 2-hydroxybenzoate; Octisalate; Octyl salicylate
Type Analytical Standard
Quantity 1 mL
Formula Weight 250.33
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less