accessibility menu, dialog, popup

UserName

OHNOO

CAS RN 118-48-9

IUPAC Name: 2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
Synonyms: 1H-3,1-benzoxazine-2,4-dione
Molecular Weight (g/mol): 163.13
Molecular Formula: C8H5NO3
InChi Key: TXJUTRJFNRYTHH-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2C(=O)O1
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)