accessibility menu, dialog, popup

UserName

CAS RN 117695-55-3

IUPAC Name: (3,5-dibromophenyl)boronic acid
Synonyms: 3,5-dibromophenylboronic acid
Molecular Weight (g/mol): 279.72
Molecular Formula: C6H5BBr2O2
InChi Key: WQBLCGDZYFKINX-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Br)=CC(Br)=C1
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results