missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D46705G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D46705G Supplier TCI America Supplier No. D46705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3,5-Dibromophenylboronic Acid (contains varying amounts of Anhydride)
CAS 117695-55-3
Molecular Formula C6H5BBr2O2
Quantity 5 g
MDL Number MFCD01075725
Synonym 3,5-dibromobenzeneboronic acid, 3,5-dibromophenyl boronic acid, boronic acid, 3,5-dibromophenyl, pubchem14108, acmc-2099ur, amtb596, 3,5-dibromobenzeneboronicacid, ksc493i3d, 3,5-dibromobenzene boronic acid
InChI Key WQBLCGDZYFKINX-UHFFFAOYSA-N
SMILES B(C1=CC(=CC(=C1)Br)Br)(O)O
IUPAC Name (3,5-dibromophenyl)boronic acid
Molecular Weight (g/mol) 279.722
PubChem CID 2734689
Formula Weight 279.72
Physical Form Crystalline Powder
Show More Show Less