accessibility menu, dialog, popup

UserName

CH3NH3COOClSOONH2

CAS RN 1172493-99-0

IUPAC Name: 4-sulfamoylbenzoyl chloride; N,N-dimethylformamide
Synonyms: 4-sulfamoylbenzoyl chloride; dimethylformamide
Molecular Weight (g/mol): 292.73
Molecular Formula: C10H13ClN2O4S
InChi Key: CRGFLVSVHAHYCI-UHFFFAOYSA-N
SMILES: CN(C)C=O.NS(=O)(=O)C1=CC=C(C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results