missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfamidobenzoyl chloride DMF complex, 95%

Catalog No. AAL1842814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1842814 25 g
AAL1842806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1842814 Supplier Thermo Scientific Chemicals Supplier No. L1842814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1172493-99-0 | C10H13ClN2O4S | 292.73 g/mol

It was used in the discovery of 2-Phenyl-3-sulfonylphenyl-indole derivatives as a new class of selective COX-2 inhibitors and in the synthesis and biological evaluation of substituted 2-sulfonyl-phenyl-3-phenyl-indoles: a new series of selective COX-2 inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used in the discovery of 2-Phenyl-3-sulfonylphenyl-indole derivatives as a new class of selective COX-2 inhibitors and in the synthesis and biological evaluation of substituted 2-sulfonyl-phenyl-3-phenyl-indoles: a new series of selective COX-2 inhibitors.

Solubility
Decomposes in water.

Notes
Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 1172493-99-0
Molecular Formula C10H13ClN2O4S
Molecular Weight (g/mol) 292.73
MDL Number MFCD02094019
InChI Key CRGFLVSVHAHYCI-UHFFFAOYSA-N
Synonym 4-sulfamidobenzoyl chloride dmf complex, 4-sulfamoylbenzoyl chloride; dimethylformamide, 4-sulphamido benzoyl chloride dmf complex, 4-aminosulfonyl benzoyl chloride dmf complex, 4-chlorocarbonyl benzenesulphonamide dmf complex
PubChem CID 44119145
SMILES CN(C)C=O.NS(=O)(=O)C1=CC=C(C=C1)C(Cl)=O

Specifications

Quantity 25 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
IUPAC Name 4-sulfamoylbenzoyl chloride; N,N-dimethylformamide
Formula Weight 292.74
Percent Purity 95%
Chemical Name or Material 4-Sulfamidobenzoyl chloride DMF complex
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only