accessibility menu, dialog, popup

UserName

CAS RN 1171974-28-9

IUPAC Name: 4,7-bis(5-octylthiophen-2-yl)-2,1,3-benzothiadiazole
Synonyms: 4,7-bis(5-octylthiophen-2-yl)-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 524.84
Molecular Formula: C30H40N2S3
InChi Key: CPPGJRXHRGQCHC-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(S1)C1=CC=C(C2=CC=C(CCCCCCCC)S2)C2=NSN=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results