Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4,7-Bis(5-n-octyl-2-thienyl)-2,1,3-benzothiadiazole 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 200 mg |
| Formula Weight | 524.84 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,7-Bis(5-n-octyl-2-thienyl)-2,1,3-benzothiadiazole |
Safety and Handling
| TSCA | No |