accessibility menu, dialog, popup

UserName

ClClOO

CAS RN 117-80-6

IUPAC Name: 2,3-dichloro-1,4-dihydronaphthalene-1,4-dione
Synonyms: dichlone
Molecular Weight (g/mol): 227.04
Molecular Formula: C10H4Cl2O2
InChi Key: SVPKNMBRVBMTLB-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)