CAS RN 1163-36-6
IUPAC Name:
hydrogen 1-[(4-chlorophenyl)methyl]-2-[(pyrrolidin-1-yl)methyl]-1H-1,3-benzodiazole chloride
Synonyms:
hydrogen clemizole chloride
Molecular Weight (g/mol):
362.3
Molecular Formula:
C19H21Cl2N3
InChi Key:
DNFMJYXRIMLMBZ-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].ClC1=CC=C(CN2C(CN3CCCC3)=NC3=CC=CC=C23)C=C1
1
–
1
of
1
results
1
Clemizole Hydrochloride 98.0+%, TCI America™
CAS: 1163-36-6 Molecular Formula: C19H21Cl2N3 Molecular Weight (g/mol): 362.30 MDL Number: MFCD00051435 InChI Key: DNFMJYXRIMLMBZ-UHFFFAOYSA-N Synonym: 1-(4-Chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)benzimidazole Hydrochloride PubChem CID: 5309446 IUPAC Name: hydrogen 1-[(4-chlorophenyl)methyl]-2-[(pyrrolidin-1-yl)methyl]-1H-1,3-benzodiazole chloride SMILES: [H+].[Cl-].ClC1=CC=C(CN2C(CN3CCCC3)=NC3=CC=CC=C23)C=C1