accessibility menu, dialog, popup

UserName

HNBr

CAS RN 1160294-93-8

IUPAC Name: N-(4-bromophenyl)-[1,1'-biphenyl]-4-amine
Synonyms: N-(4-bromophenyl)-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 324.22
Molecular Formula: C18H14BrN
InChi Key: RDSXVQDXXSPFHG-UHFFFAOYSA-N
SMILES: BrC1=CC=C(NC2=CC=C(C=C2)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results