missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromophenyl)-4-biphenylamine 98.0+%, TCI America™

Catalog No. B47535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47535G 5 g
B47531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47535G Supplier TCI America Supplier No. B47535G

Chemical Identifiers

CAS 1160294-93-8
Molecular Formula C18H14BrN
Molecular Weight (g/mol) 324.221
MDL Number MFCD20486764
InChI Key RDSXVQDXXSPFHG-UHFFFAOYSA-N
Synonym 4-(4-Bromoanilino)biphenyl
PubChem CID 57746339
IUPAC Name N-(4-bromophenyl)-4-phenylaniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=C(C=C3)Br

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 5 g
Formula Weight 324.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Bromophenyl)-4-biphenylamine