CAS RN 1154-59-2
IUPAC Name:
3,5-dichloro-N-(3,4-dichlorophenyl)-2-hydroxybenzamide
Synonyms:
Tcsa TCSA
Molecular Weight (g/mol):
351
Molecular Formula:
C13H7Cl4NO2
InChi Key:
SJQBHPJLLIJASD-UHFFFAOYSA-N
SMILES:
OC1=C(Cl)C=C(Cl)C=C1C(=O)NC1=CC=C(Cl)C(Cl)=C1
1
–
1
of
1
results
1
Promotions Available
3,3',4',5-Tetrachlorosalicylanilide, 97%
3,3',4',5-Tetrachlorosalicylanilide, 97%
CAS: 1154-59-2 Molecular Formula: C13H7Cl4NO2 Molecular Weight (g/mol): 351.01 MDL Number: MFCD00041745 InChI Key: SJQBHPJLLIJASD-UHFFFAOYSA-N Synonym: tcsa,3,3',4',5-tetrachlorosalicylanilide,3,5-dichloro-n-3,4-dichlorophenyl-2-hydroxybenzamide,3,5,3',4'-tetrachlorosalicylanilide,irgasan bs-200,3,5-dichlorosalicyl 3,4-dichloroanilide,caswell no. 833,3,5-dichlorosalicyl-3,4-dichloroanilide,benzamide, 3,5-dichloro-n-3,4-dichlorophenyl-2-hydroxy,unii-hne676755i PubChem CID: 14385 ChEBI: CHEBI:188648 IUPAC Name: 3,5-dichloro-N-(3,4-dichlorophenyl)-2-hydroxybenzamide SMILES: C1=CC(=C(C=C1NC(=O)C2=CC(=CC(=C2O)Cl)Cl)Cl)Cl