CAS RN 1152-62-1
CAS RN 1152-62-1
IUPAC Name:
2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Synonyms:
2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Molecular Weight (g/mol):
283.34
Molecular Formula:
C13H17NO4S
InChi Key:
FPKHNNQXKZMOJJ-UHFFFAOYNA-N
SMILES:
CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
N-Carbobenzoxy-L-methionine 98.0+%, TCI Americaâ„¢
CAS: 1152-62-1 Molecular Formula: C13H17NO4S Molecular Weight (g/mol): 283.34 MDL Number: MFCD00065124 InChI Key: FPKHNNQXKZMOJJ-UHFFFAOYNA-N Synonym: z-met-oh,n-cbz-l-methionine,cbz-l-methionine,cbz-met-oh,z-l-methionine,n-carbobenzyloxy-l-methionine,n-carbobenzoxy-l-methionine,benzyloxycarbonyl-l-methionine,n-benzyloxy carbonyl-l-methionine,n-benzyloxycarbonyl-l-methionine PubChem CID: 1550857 IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid SMILES: CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O