missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-méthionine 98,0+%, TCI America™

Catalog No. C07375G
Change view
Click to view available options
Quantité:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
C07375G 5 g
C073725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07375G Supplier TCI America Supplier No. C07375G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1152-62-1
Formule moléculaire C13H17NO4S
Poids moléculaire (g/mol) 283.34
Numéro MDL MFCD00065124
Clé InChI FPKHNNQXKZMOJJ-UHFFFAOYNA-N
Synonyme z-met-oh, n-cbz-l-methionine, cbz-l-methionine, cbz-met-oh, z-l-methionine, n-carbobenzyloxy-l-methionine, n-carbobenzoxy-l-methionine, benzyloxycarbonyl-l-methionine, n-benzyloxy carbonyl-l-methionine, n-benzyloxycarbonyl-l-methionine
PubChem CID 1550857
Nom de l’IUPAC 2-{[(benzyloxy)carbonyl]amino}-4-(méthylsulfanyl)acide butanoïque
SOURIRES CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Nom ou substance chimique N-Carbobenzoxy-L-methionine
Point de fusion 68 °C
Couleur Blanc-jaune
Quantité 5 g
Poids de formule 283.34
Pourcentage de pureté ≥98,0% (T)
Forme physique Poudre cristalline