accessibility menu, dialog, popup

UserName

CH3OOONH3CCH3CH3CH3

CAS RN 114768-72-8

IUPAC Name: 5-acetyl-10,10,16,16-tetramethyl-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Synonyms: 5-acetyl-10,10,16,16-tetramethyl-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Molecular Weight (g/mol): 339.44
Molecular Formula: C21H25NO3
InChi Key: NGTDVJOJDMQAMI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=CC3=C4N(CCC3(C)C)CCC(C)(C)C4=C2OC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results