missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 521T 98.0+%, TCI America™

Catalog No. C3046200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3046200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C3046200MG Supplier TCI America Supplier No. C3046200MG

Chemical Identifiers

CAS 114768-72-8
Molecular Formula C21H25NO3
Molecular Weight (g/mol) 339.435
InChI Key NGTDVJOJDMQAMI-UHFFFAOYSA-N
Synonym 10-Acetyl-2,3,6,7-tetrahydro-1,1,7,7-tetramethyl-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one
PubChem CID 14365279
SMILES CC(=O)C1=CC2=CC3=C4C(=C2OC1=O)C(CCN4CCC3(C)C)(C)C

Specifications

Quantity 200 mg
Formula Weight 339.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 521T