accessibility menu, dialog, popup

UserName

CAS RN 1143576-84-4

IUPAC Name: 4,4,5,5-tetramethyl-2-[4-(10-phenylanthracen-9-yl)phenyl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[4-(10-phenylanthracen-9-yl)phenyl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 456.39
Molecular Formula: C32H29BO2
InChi Key: MLLYTNZYCPAKPU-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results