missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-[4-(10-phenylanthracen-9-yl)phenyl]-1,3,2-dioxaborolane 95.0+%, TCI America™

Catalog No. T3276200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3276200MG 200 mg
T32761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3276200MG Supplier TCI America Supplier No. T3276200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1143576-84-4
Molecular Formula C32H29BO2
Molecular Weight (g/mol) 456.392
InChI Key MLLYTNZYCPAKPU-UHFFFAOYSA-N
Synonym 9-Phenyl-10-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]anthracene, 4-(10-Phenylanthracen-9-yl)phenylboronic Acid Pinacol Ester
PubChem CID 57654767
IUPAC Name 4,4,5,5-tetramethyl-2-[4-(10-phenylanthracen-9-yl)phenyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=CC=CC=C6

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 456.39
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,5,5-Tetramethyl-2-[4-(10-phenylanthracen-9-yl)phenyl]-1,3,2-dioxaborolane