accessibility menu, dialog, popup

UserName

CHOOHCOHC

CAS RN 114233-80-6

IUPAC Name: 1,3,5-tris(prop-2-yn-1-yloxy)benzene
Synonyms: 1,3,5-tris(prop-2-yn-1-yloxy)benzene
Molecular Weight (g/mol): 240.26
Molecular Formula: C15H12O3
InChi Key: WVZIFQJSKQJHDU-UHFFFAOYSA-N
SMILES: C#CCOC1=CC(OCC#C)=CC(OCC#C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results