missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(2-propynyloxy)benzene 98.0+%, TCI America™

Catalog No. T3135200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3135200MG 200 mg
T31351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3135200MG Supplier TCI America Supplier No. T3135200MG

Chemical Identifiers

CAS 114233-80-6
Molecular Formula C15H12O3
Molecular Weight (g/mol) 240.258
InChI Key WVZIFQJSKQJHDU-UHFFFAOYSA-N
Synonym 1,3,5-Tris(propargyloxy)benzene
PubChem CID 11630083
IUPAC Name 1,3,5-tris(prop-2-ynoxy)benzene
SMILES C#CCOC1=CC(=CC(=C1)OCC#C)OCC#C

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 200 mg
Formula Weight 240.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(2-propynyloxy)benzene