accessibility menu, dialog, popup

UserName

NN

CAS RN 113682-56-7

IUPAC Name: 4-[4-(pyridin-4-yl)phenyl]pyridine
Synonyms: 4-[4-(pyridin-4-yl)phenyl]pyridine
Molecular Weight (g/mol): 232.29
Molecular Formula: C16H12N2
InChi Key: MAWKLXRVKVOYLR-UHFFFAOYSA-N
SMILES: C1=CC(=CC=N1)C1=CC=C(C=C1)C1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results