missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Di(4-pyridyl)benzene 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 193°C |
| Color | White-Yellow |
| Quantity | 200 mg |
| Formula Weight | 232.29 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,4-Di(4-pyridyl)benzene |
Safety and Handling
| TSCA | No |