man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 113421-98-0

NH2OFFFF

CAS RN 113421-98-0

IUPAC Name: 4-fluoro-3-(trifluoromethoxy)aniline
Synonyms: 4-fluoro-3-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 195.12
Molecular Formula: C7H5F4NO
InChi Key: JSMGZPOPIVVIRZ-UHFFFAOYSA-N
SMILES: NC1=CC(OC(F)(F)F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results