accessibility menu, dialog, popup

UserName

H3COOHOHO

CAS RN 1131-94-8

IUPAC Name: 2-(3-hydroxy-4-methoxyphenyl)acetic acid
Synonyms: isohomovanillic acid isohomovanillate homoisovanillic acid homoisovanillate homo-iso-vanillic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: BWXLCOBSWMQCGP-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(O)=O)C=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results