missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. H14555G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14555G 5 g
H14551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14555G Supplier TCI America Supplier No. H14555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1131-94-8
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00016829
InChI Key BWXLCOBSWMQCGP-UHFFFAOYSA-N
Synonym Homoisovanillic Acid
PubChem CID 160562
ChEBI CHEBI:70818
IUPAC Name 2-(3-hydroxy-4-methoxyphenyl)acetic acid
SMILES COC1=C(C=C(C=C1)CC(=O)O)O

Specifications

Melting Point 131°C
Color Yellow
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4-methoxyphenylacetic Acid