accessibility menu, dialog, popup

UserName

CH3NOHNNNNONH3COCH3

CAS RN 1124329-14-1

IUPAC Name: 9-cyclopentyl-2-({2-methoxy-4-[(1-methylpiperidin-4-yl)oxy]phenyl}amino)-7-methyl-8,9-dihydro-7H-purin-8-one
Synonyms: 9-cyclopentyl-2-({2-methoxy-4-[(1-methylpiperidin-4-yl)oxy]phenyl}amino)-7-methylpurin-8-one
Molecular Weight (g/mol): 452.56
Molecular Formula: C24H32N6O3
InChi Key: YUKWVHPTFRQHMF-UHFFFAOYSA-N
SMILES: COC1=C(NC2=NC=C3N(C)C(=O)N(C4CCCC4)C3=N2)C=CC(OC2CCN(C)CC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results