accessibility menu, dialog, popup

UserName

H3CNH2O

CAS RN 1120-16-7

IUPAC Name: dodecanamide
Synonyms: lauramide
Molecular Weight (g/mol): 199.34
Molecular Formula: C12H25NO
InChi Key: ILRSCQWREDREME-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(N)=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results