missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lauramide 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 99°C |
| Color | White-Yellow |
| Boiling Point | 200°C |
| Quantity | 500 g |
| Formula Weight | 199.34 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Lauramide |
Safety and Handling
| EINECSNumber | (2)-0824 |
| TSCA | Yes |