accessibility menu, dialog, popup

UserName

CH3NH3COHO

CAS RN 1118-68-9

IUPAC Name: 2-(dimethylamino)acetic acid
Synonyms: dimethylglycine dimethylglycine, calcium salt monopotassium salt dimethylglycine dimethylglycine, monopotassium salt calcium salt dimethylglycine sodium salt dimethylglycine dimethylglycine, sodium salt
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: FFDGPVCHZBVARC-UHFFFAOYSA-N
SMILES: CN(C)CC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)