missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N-Dimethylglycine, 97%

Catalog No. AC345200250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC345200250 25 g Glass bottle
AC345200050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC345200250 Supplier Thermo Scientific Chemicals Supplier No. 345200250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1118-68-9 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1118-68-9
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00004283
InChI Key FFDGPVCHZBVARC-UHFFFAOYSA-N
Synonym n,n-dimethylglycine, dimethylglycine, 2-dimethylamino acetic acid, dimethylamino acetic acid, n,n-dimethylaminoacetic acid, glycine, n,n-dimethyl, n-methylsarcosine, n,n-dimethyl glycine, unii-7797m4cppa, dmg
PubChem CID 673
ChEBI CHEBI:17724
IUPAC Name 2-(dimethylamino)acetic acid
SMILES CN(C)CC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 175.0°C to 181.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)2NCH2COOH
Quantity 25 g
Merck Index 15, 3269
Solubility Information Solubility in water: soluble
Formula Weight 103.12
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Dimethylglycine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves.
EINECSNumber 214-267-8
RTECSNumber MB9865000
TSCA TSCA

RUO – Research Use Only