accessibility menu, dialog, popup

UserName

H3C(S)O(R)(S)OHOHNNNHO

CAS RN 111358-88-4

IUPAC Name: (15S,16S,18R)-16-hydroxy-16-(hydroxymethyl)-15-methyl-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8,10,12,20(25),21,23-nonaen-3-one
Synonyms: lestaurtinib
Molecular Weight (g/mol): 439.47
Molecular Formula: C26H21N3O4
InChi Key: UIARLYUEJFELEN-LROUJFHJSA-N
SMILES: C[C@]12O[C@H](C[C@]1(O)CO)N1C3=C(C=CC=C3)C3=C1C1=C(C4=C3C(=O)NC4)C3=CC=CC=C3N21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results