accessibility menu, dialog, popup

UserName

ClH3CNCH3H3CH3C

CAS RN 1112-67-0

IUPAC Name: tetrabutylazanium chloride
Synonyms: tetrabutylammonium chloride tetra-N-butylammonium chloride
Molecular Weight (g/mol): 277.92
Molecular Formula: C16H36ClN
InChi Key: NHGXDBSUJJNIRV-UHFFFAOYSA-M
SMILES: [Cl-].CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (4)
  • (1)
  • (5)
Quantity 
  • (2)
  • (4)
  • (3)
  • (1)
Percent Purity 
  • (5)
  • (4)
Physical Form 
  • (5)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (5)
  • (4)
  • (4)
  • (1)
  • (5)
  • (2)
  • (4)
  • (3)
  • (1)
  • (5)
  • (4)
  • (5)
  • (4)
  • (4)