missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Chloride 98.0+%, TCI America™

Catalog No. I03665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03665G 5 g
I036625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03665G Supplier TCI America Supplier No. I03665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1112-67-0
Molecular Formula C16H36ClN
Molecular Weight (g/mol) 277.921
MDL Number MFCD00011635
InChI Key NHGXDBSUJJNIRV-UHFFFAOYSA-M
Synonym tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride
PubChem CID 70681
ChEBI CHEBI:51988
IUPAC Name tetrabutylazanium;chloride
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[Cl-]

Specifications

Melting Point 70°C
Color White
Quantity 5 g
Formula Weight 277.92
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Chloride [Reagent for Ion-Pair Chromatography]
EINECSNumber (2)-0186
TSCA Yes