accessibility menu, dialog, popup

UserName

H3CBr

CAS RN 111-83-1

IUPAC Name: 1-bromooctane
Synonyms: 1-bromooctane
Molecular Weight (g/mol): 193.13
Molecular Formula: C8H17Br
InChi Key: VMKOFRJSULQZRM-UHFFFAOYSA-N
SMILES: CCCCCCCCBr
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)