missing translation for 'onlineSavingsMsg'
Learn More

Triglyceride Mix C2 - C10, Neat, MilliporeSigma™ Supelco™

Catalog No. 111012476
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012476 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012476 Supplier Merck Emd Millipore Supplier No. 178101AMPS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Ampule
CAS 111-83-1
Physical Form Liquid
Synonym Lipid standards: triglyceride mixtures
Quantity 100 mg
Grade Analytical Standard