missing translation for 'onlineSavingsMsg'
Learn More

1-Bromooctane 98.0+%, TCI America™

Catalog No. B0626500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0626500ML 500 mL
B062625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0626500ML Supplier TCI America Supplier No. B0626500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-83-1
Molecular Formula C8H17Br
Molecular Weight (g/mol) 193.13
MDL Number MFCD00000276
InChI Key VMKOFRJSULQZRM-UHFFFAOYSA-N
Synonym n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo
PubChem CID 8140
IUPAC Name 1-bromooctane
SMILES CCCCCCCCBr

Specifications

Color Colorless
Boiling Point 201°C
Quantity 500 mL
UN Number 3082
Formula Weight 193.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromooctane
EINECSNumber (2)-0067
RTECSNumber RG8575000
TSCA Yes