accessibility menu, dialog, popup

UserName

H3CNHHNCH3

CAS RN 111-74-0

IUPAC Name: ethyl[2-(ethylamino)ethyl]amine
Synonyms: N,N'-diethylethylenediamine
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: CJKRXEBLWJVYJD-UHFFFAOYSA-N
SMILES: CCNCCNCC
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)