missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diethylethylenediamine, 95%

Catalog No. AC155520050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155520050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC155520050 Supplier Thermo Scientific Chemicals Supplier No. 155520050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-74-0 | C6H16N2 | 116.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-74-0
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
MDL Number MFCD00009033
InChI Key CJKRXEBLWJVYJD-UHFFFAOYSA-N
Synonym n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide
PubChem CID 67105
ChEBI CHEBI:182290
IUPAC Name N,N'-diethylethane-1,2-diamine
SMILES CCNCCNCC

Specifications

Density 0.8110g/mL
Boiling Point 152.0°C to 154.0°C
Flash Point 33°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4326
Linear Formula C2H5NHCH2CH2NHC2H5
Quantity 5 g
Beilstein 04,251
Specific Gravity 0.811
Solubility Information Solubility in water: miscible.
Formula Weight 116.21
Percent Purity 95%
Chemical Name or Material N, N'-Diethylethylenediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 203-902-4

RUO – Research Use Only