accessibility menu, dialog, popup

UserName

NN

CAS RN 111-69-3

IUPAC Name: hexanedinitrile
Synonyms: adiponitrile
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: BTGRAWJCKBQKAO-UHFFFAOYSA-N
SMILES: N#CCCCCC#N
Boiling Point 
  • (6)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)