missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dicyanobutane, 99%, Thermo Scientific Chemicals

Catalog No. AC113831000
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113831000 100 mL Glass bottle
AC113830010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC113831000 Supplier Thermo Scientific Chemicals Supplier No. 113831000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-69-3 | Molecular Formula: C6H8N2 | Molecular Weight: 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-69-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00001975
InChI Key BTGRAWJCKBQKAO-UHFFFAOYSA-N
Synonym adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril
PubChem CID 8128
IUPAC Name hexanedinitrile
SMILES C(CCC#N)CC#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 1.0°C to 3.0°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 295.0°C
Flash Point 163°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4370 to 1.4390
Linear Formula NC(CH2)4CN
Quantity 100 mL
Beilstein 02, 653
Specific Gravity 0.95
Merck Index 15, 152
Solubility Information Solubility in water: 90g/L (20°C). Other solubilities: soluble in alcohol and chloroform
Viscosity 5.8 mPa.s (30°C)
Formula Weight 108.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 4-Dicyanobutane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and
EINECSNumber 203-896-3
RTECSNumber AV2625000
TSCA TSCA

RUO – Research Use Only