accessibility menu, dialog, popup

UserName

CAS RN 111-29-5

IUPAC Name: pentane-1,5-diol
Synonyms: 1,5-pentanediol
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: ALQSHHUCVQOPAS-UHFFFAOYSA-N
SMILES: OCCCCCO
Boiling Point 
  • (2)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (2)
  • (3)
  • (4)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)