accessibility menu, dialog, popup

UserName

CAS RN 111-11-5

IUPAC Name: methyl octanoate
Synonyms: methyl caprylate
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: JGHZJRVDZXSNKQ-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OC
Boiling Point 
  • (3)
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (7)
  • (3)
  • (2)
  • (5)
  • (5)
  • (5)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)