missing translation for 'onlineSavingsMsg'
Learn More

Octanoate de méthyle n-octanoate 99,5+%, TCI America™

Catalog No. S03065ML
Change view
Click to view available options
Quantité:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
S03065ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03065ML Supplier TCI America Supplier No. S03065ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-11-5
Formule moléculaire C9H18O2
Poids moléculaire (g/mol) 158.241
Numéro MDL MFCD00009551
Clé InChI JGHZJRVDZXSNKQ-UHFFFAOYSA-N
Synonyme methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
PubChem CID 8091
ChEBI CHEBI:87432
Nom de l’IUPAC Octanoate de méthyle
SOURIRES CCCCCCCC(=O)OC

Specifications

Nom ou substance chimique Methyl n-Octanoate [Standard Material for GC]
Couleur Incolore
Point d’ébullition 193 °C
Quantité 5 mL
Poids de formule 158.24
Pourcentage de pureté ≥99,5% (GC)
Forme physique Liquide
EINECSNumber (2)-0782
RTECSNumber RH0778000
TSCA Oui