accessibility menu, dialog, popup

UserName

H3CCH3O

CAS RN 110-43-0

IUPAC Name: heptan-2-one
Synonyms: 2-heptanone fema number 2544 methyl-amyl-cetone amyl-methyl-cetone ketone C-7 2-heptanone (natural)
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: CATSNJVOTSVZJV-UHFFFAOYSA-N
SMILES: CCCCCC(C)=O
Percent Purity 
  • (2)
  • (1)
  • (4)
  • (2)
Boiling Point 
  • (2)
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (6)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (4)